Would you like to log in again?
Visual Scientifics Diffraction
Supplier: EISCO SCIENTIFIC LLC
PTDSE
470212-492EA 54.3 USD InStock
470212-492
Visual Scientifics Diffraction
Educational Materials Physics Educational Materials Optics Learning Activities
Use this apparatus to make your own spectroscope! A simple, yet effective device to explore the wave/particle duality of light.
  • Versatile, precise, and easy to assemble
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
  • 10+ page instruction book included
The concept of wave and particle duality is typically difficult to understand and construct without and expensive optical bench, but using the Visual Scientifics system, it is fast and easy to set up and explore. Students will derive and experimentally prove the relationship y=(mL)/d. The included instruction booklet provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts. Every module includes comprehensive instructions to assist with lesson planning, student activities, and experimental procedures.
Ordering information: Visual Scientifics Base (470148-822) and Backboard (470135-558) required, but sold separately.
Delivery information: Included in this kit are a mounted laser, support posts, 3 different diffraction gratings, and slide holder. Recommended accessories: video camera, graphical analysis software, and a ruler.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty