Would you like to log in again?
Ink Chromatography and Forensics STEM Kit
Supplier: United Scientific Supplies
AISCRKIT
470213-358EA 99.45 USD InStock
470213-358
Ink Chromatography and Forensics STEM Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Students solve the case with ink chromatography and STEM.
  • Hands-On STEM Activity
  • Model and Inquiry Activities
  • Teacher and Student Guides Included
  • Material Accommodates up to 40 Students
  • Covered Concepts Include Data Analysis, Chromatographs, and More
This kit introduces students to ink chromatography and its application in solving forensics cases. They gain experience in creating ink databases, identifying ‘ink tags’ to ‘date’ various inks and then use them, as professional document examiners do, in solving two mysteries involving document inks—an apparent suicide note and a forgery case.
Covered concepts include data analysis, chromatographs, inks and their composition-mixtures, compounds, mixtures, molecules, solubility, affinity, polar substances, Rf values, capillary action, light energy, fluorescence, and hypothesis testing.
Ordering information: This kit includes materials for three detailed activities (with ‘model’ and ‘inquiry’ investigational paths—a total of four investigational activities), which can accommodate up to 40 students. It also contains a DVD with PDFs of teacher and student guides and other digital content.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty