Would you like to log in again?
The Chemistry of Invisible Inks STEM Kit
Supplier: United Scientific Supplies
AISINKIT
470213-360EA 96.75 USD InStock
470213-360
The Chemistry of Invisible Inks STEM Kit
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Explore How Invisible Inks Are Made
  • Inquiry-based hands-on laboratory
  • Guided and independent investigations
  • STEM Integration
This kit is a unique teaching tool for introducing your students to the fascinating chemistry and physics behind invisible inks, i.e. colorless substances usable for writing that can be made to turn to color using a chemical or physical process. Students investigate how organic fluids and chemical reagents can be used as invisible inks. Relying on historical example, students create various invisible inks (and security) documents for other student groups to analyze and decode! Includes materials for two detailed activities (each with a ‘model’ and ‘inquiry’ investigation paths – a total of four investigational activities). Covered concepts include: data analysis, chemical & physical processes, elements, mixtures & compounds, pH indicators, transmitted vs. reflected light, the periodic table, fluorescent compounds, types of chemical reactions, and hypothesis testing.
The kit includes materials for two detailed activities (each with a ‘model’ and ‘inquiry’ investigation paths for a total of four investigational activities). Covered concepts include data analysis, chemical and physical processes, elements, mixtures and compounds, pH indicators, transmitted versus reflected light, the periodic table, fluorescent compounds, types of chemical reactions, and hypothesis testing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty