Would you like to log in again?
Gear Demonstration and Transformation of Energy
Supplier: LAB CONNECTIONS SE
10058
470213-752EA 258.99 USD InStock
470213-752
Gear Demonstration and Transformation of Energy
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Gear Demonstration & Transformation of Energy
An excellent robust working model. The model clearly shows how various, most widely used Gears Work and also demonstrates how Electrical Energy can be converted into mechanical energy. The unit consists of a mounted motor which is connected by a belt and pulley to a train of different kinds of Gears. The various kinds of gears demonstrated are, Straight Bevel Gears, Worm Gears and Spur Gears. The transformation of Energy / Motion is demonstrated by Pulley Belt working, Gears connected by Chain working (bicycle) and transformation of energy to a wheel. Base: 12 inch(300mm) x 8 inch(200mm) x 0.60 inch(15mm). Height of unit, taking into account the tallest part 5inch(125mm). Gears vary in diameter, smallest being 1 inch(25mm), biggest being 3.75inch(95mm). The motor works on 6v dc, battery holder for 4 D cell batteries supplied. Batteries not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty