Would you like to log in again?
Torsion of Metal Rods Apparatus
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH10613T
470218-906EA 198.85 USD InStock
470218-906
Torsion of Metal Rods Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Torsion of Metal Rods Apparatus shows students the affect of a torque on a metal rod.
  • Two Graduated Scales
  • Heavy-Metal Pulleys
  • Connected Rods
  • Two Pointers
  • For demonstrating the Affect of a Torque on a Metal Rod
Connected rods hold two heavy-metal brackets together. A heavy-metal pulley, moving in ball bearings, is mounted on one bracket and is fitted with a three-jaw chuck to hold one end of the test rod. The other end is held in another chuck fixed to the other bracket. Two graduated scales are mounted on the connecting rod, and two pointers can be clamped in any desired position on the test rod with one brass and one steel test rod.
Hanging masses and C clamps are required but sold separately. For best results, use masses heavier than 1 kg.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty