Would you like to log in again?
Cartesian Diver Set
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH10634/D PH10634
470218-910EA 19.5 USD InStock
470218-910 470218-908
Cartesian Diver Set
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This diver set expresses the effects of air pressure and buoyancy.
  • Topics Include Transmission of Pressure by Liquid, Compressibility of Gases, and Archimedes’ Principle
  • Set Also Expresses Effects of Air Pressure and Buoyancy
  • Instructions Included
This apparatus is ideal for showing the transmission of pressure by a liquid, the compressibility of gases, and Archimedes’ Principle.
Instructions are included.
Ordering information: This kit includes a Cartesian diver, a 25 x 4 cm dia. glass hydrometer cylinder, a rubber diaphragm, and instructions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty