Would you like to log in again?
Specific Heat Demonstration Set
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH20202 PH20202S
470218-914PK 32.6 USD InStock
470218-914 470218-912
Specific Heat Demonstration Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Visually illustrate the difference in specific heat.
  • Illustrate the different heat capacities of metal
A dramatic way to illustrate the different heat capacities of metal, this slide features a slanted panel with four grooves to hold four different metals. Simply place a sheet of bee's wax in the front of the plane and submerge the metal samples in hot water. When the metals are placed on the grooves simultaneously they will melt and sink through to different levels based on their specific heat. Includes four metals cylinders: aluminum, copper, lead, and tin; support stand, two pieces of bees wax, and instructions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty