Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic Acid

Supplier: ALDON CORP SE
AA0010-1L AA0008-500MLR AA0005-2.5L AA0007-500ML AA0005-PK/2R AD-AA0005P-PK/6 AD-AA0005P-2.5L AA0004-200ML AA0003-500ML AA0005-100MLR AA0005-CONC AA0005-250ML AA0005-500MLR AA0015-1L AD-AA0009-3.8L AA0008-2.5LR
470225-684EA 11.45 USD InStock
470225-684 470300-042 470225-820 470219-202 470300-030 470300-040 470300-028 470300-038 470300-048 470300-036 470300-046 470300-034 470300-044 470300-032
Acetic Acid
Chemicals for Science Education
CAS Number: 64-19-7
Formula: CH3COOH
Density: 1.049 g/ml
Boiling Point: 118.1°C
Freezing Point: 16.7°C

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: The item is corrosive, flammable and toxic.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty