Would you like to log in again?
Analog Sensor Extension Cable
Supplier: VERNIER SOFTWARE & TECH SE
EXT-BTA
470219-276EA 11.99 USD InStock
470219-276
Analog Sensor Extension Cable
Cables
Most of the Vernier sensors have cables that are 1.5 m long.
There are times, however, that users may wish to have a longer cable. The Analog Sensor Extension Cable can be used with any Vernier sensor that connects to an analog channel labeled CH1, CH2, CH3, or CH4. Each cable is 2 m in length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty