Would you like to log in again?
Mini to Standard USB Adapter
Supplier: VERNIER SOFTWARE & TECH SE
MINI-USB
470219-302EA 21.99 USD InStock
470219-302
Mini to Standard USB Adapter
Cables
This adapter allows an EasyTemp or EasyLink to be connected to a computer for data collection using Logger Pro or Logger Lite software. The adapter has a USB Mini-A receptacle that connects to the EasyTemp or EasyLink and a standard-A plug that connects to the computer. It can also be used with the USB cable that comes with the CBR 2.
Connector 1: USB Mini Type A Receptacle
Connector 2: USB Standard Type A Plug
Note: Most consumer electronic devices use a USB Mini Type B plug, and will NOT WORK with this adapter. Our adapter only works for devices with USB Mini Type A plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty