Would you like to log in again?
Lenz's Law
Supplier: EISCO SCIENTIFIC LLC
LZLWKIT
470219-408EA 70.25 USD InStock
470219-408
Lenz's Law
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Lenz's Law Kit investigates how much force an induced magnetic field can create.
  • Kit Also Explores Faraday's Law, Magnetism, and Eddy Currents
  • Clear Plastic Frame Holds an Aluminum Tube and a Plastic Tube
  • Frame Measures Around 12.5" in Height
  • Magnets and Dynamometer 2.5N/250 g Included
  • Experiment Guide Included
It provides the tools necessary to discover concepts involving Faraday's Law, magnetism, and eddy currents in addition to Lenz's Law.
The pipes of differing materials demonstrate the magnet braking force in action as two magnets are simultaneously dropped through two different tubes. One tube is aluminum, and other is plastic. A dynamometer attaches to suspend the Lenz's Law apparatus, so students can quantitatively measure the additional force as the magnet drops through the conductive tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty