Would you like to log in again?
Spring Large - Compression
Supplier: EISCO SCIENTIFIC LLC
PH0115C
470219-412EA 12.1 USD InStock
470219-412
Spring Large - Compression
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Excellent demonstration of compression for hooke's law. Approximately 300mm long with a 10mm diameter and wire of 19 SWG.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty