Would you like to log in again?
Optical Prisms
Supplier: EISCO SCIENTIFIC LLC
PH0562A PH0562C PH0562B PH0558C PH0558A PH0558B PH0560A PH0560C PH0560B
470219-456EA 16.6 USD InStock
470219-456 470219-446 470219-458 470219-462 470219-452 470219-454 470219-460 470219-450 470219-448
Optical Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Right angle and equilateral prisms are available in a variety of forms.
  • For Investigating Refraction and Creating Rainbows
  • Non-Optically Worked
  • Polished Faces
  • Beveled Edges
  • Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty