Would you like to log in again?
Optical Refraction Components
Supplier: EISCO SCIENTIFIC LLC
PH0570A PH0568B PH0577B PH0568A PH0570B PH0566C PH0569B PH0566A PH0566E PH0570C PH0570D PH0566B PH0568D PH0577A PH0568C PH0569A PH0570E
470219-480EA 9.35 USD InStock
470219-480 470219-470 470219-492 470219-482 470219-472 470219-494 470219-490 470219-488 470219-466 470219-478 470219-468 470219-484 470219-474 470219-496 470219-486 470219-464 470219-476
Optical Refraction Components
Educational Materials Physics Educational Materials Optics Learning Activities
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
  • Students Measure the Refraction of Light
  • Refraction Blocks, Prisms, and Lenses
  • Optically Worked Surfaces
  • Bubble-Free Glass
  • Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty