Would you like to log in again?
CENCO® Spectrometer Prisms
Supplier: EISCO SCIENTIFIC LLC
PH0583D PH0583B PH0583A PH0583E PH0583C
470219-500EA 45.55 USD InStock
470219-500 470219-502 470219-498 470219-504 470219-506
CENCO® Spectrometer Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrometer prisms offer a different index of refraction for every need.
  • Different Indices of Refractions for Every Need
  • Glass Material
  • Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty