Would you like to log in again?
Tangent Galvanometer
Supplier: EISCO SCIENTIFIC LLC
PH0812
470219-512EA 127.05 USD InStock
470219-512
Tangent Galvanometer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Measure an electric current by determining the magnitude and direction of the horizontal component of Earth’s magnetic field. Set the compass needle parallel to the coil, pass an unknown current through, and measure the angle of deflection. Features copper magnet wire, adjustable potentiometer, compass, and non-magnetic stand. Required but not included is one D battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty