Would you like to log in again?
Oersted's Apparatus
Supplier: EISCO SCIENTIFIC LLC
PH0838
470219-514EA 64.4 USD InStock
470219-514
Oersted's Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Oersted's Apparatus demonstrates the magnetic field created by a current.
  • Machine Shows Action of Electric Current on a Magnet
  • Cobalt-Steel Needle
  • Agate Bearing
  • Plastic Base
  • Rectangular Frame Fitted with Terminals
In other words, it shows the action of electric current on a magnet.
A cobalt-steel needle (75 mm long) with agate bearing supported by a pivot is mounted on a plastic base and surrounded by a rectangular frame fitted with terminals. A power supply is required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty