Would you like to log in again?
Demonstration Transformer Kit
Supplier: EISCO SCIENTIFIC LLC
PH0849GT PH0849AK1
470219-518EA 695.95 USD InStock
470219-518 470219-516
Demonstration Transformer Kit
Transformers
The Demonstration Transformer Kit is a safe and convenient system for AC and DC experiments up to 0.5 amps.
  • For AC and DC Experiments up to 0.5A
  • Large-Scale Transformer
  • Ideal for the Classroom
  • Intermediary Outputs
  • Safe and Convenient
The large-scale transformer is ideal for the classroom.
The coils are fitted with intermediary outputs that provide a variety of transformation ratios.
Accessories information: The accessory kit demonstrates how step down transformers can produce currents that are large enough to weld metals or boil water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty