Would you like to log in again?
Conductors; Set of Three
Supplier: EISCO SCIENTIFIC LLC
PH0880
470219-522PK 81.15 USD InStock
470219-522
Conductors; Set of Three
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Made of brass, interchangeable on pillars, fitted with insulators of ebonite on base. Spheres 5cm diameter, Cylinder 12.5 x 5cm diameter, Cone 10 x 5cm length x diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty