Would you like to log in again?
Needle Electroscope, Large
Supplier: EISCO SCIENTIFIC LLC
PH0916
470219-526EA 81.65 USD InStock
470219-526
Needle Electroscope, Large
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This extra large electroscope is ideal for classroom demonstrations of electrostatic principles. A lightweight metal vane is used as a pointer in the electroscope. This lightweight metal blade deflects when the electroscope plate is charged. A transparent scale is provided to measure the angle of the deflection. Front glass window slides up to allow insertion of ionizing material in chamber.
This electroscope comes with two accessories: an electrophorus disc and a spherical electrode.
Assembled, the electroscope measures 9.75 x 8 x 4.75".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty