Would you like to log in again?
Acrylic Pipe Demonstration
Supplier: EISCO SCIENTIFIC LLC
PH100LCB
470219-530EA 19.25 USD InStock
470219-530
Acrylic Pipe Demonstration
Educational Materials Physics Educational Materials Optics Learning Activities
Show students how light reflects inside an acrylic pipe.
  • Transmits light like fiber optic cable
This hard acrylic 1/2" diameter light pipe is 2 feet of pipe coiled up at one end to demonstrate light transmission end to end. This light pipe is approx. 9" wide due to the coil in it.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty