Would you like to log in again?
Bottle Rocket Launcher
Supplier: EISCO SCIENTIFIC LLC
PH1120143
470219-532EA 72.7 USD InStock
470219-532
Bottle Rocket Launcher
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The Bottle Rocket Launcher uses the power of compressed air and water for liftoff.
  • Launcher Can Fire Bottles over 100 ft. into the Air
  • Corded Release Mechanism for Remote Launches Included
  • Compressed Air and Water Function as Fuel
  • Pumps Pressurize Bottles
When attached to a sturdy base, the launcher can fire 2 L plastic bottles over 100 feet in the air.
Using water as liquid fuel, pressurize the bottle with a bicycle pump (not included). Remotely launch from a safe distance with the corded release mechanism.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty