Would you like to log in again?
Decade Resistance Box, 6 Decade
Supplier: EISCO SCIENTIFIC LLC
PH1133AN8
470219-538EA 107.65 USD InStock
470219-538
Decade Resistance Box, 6 Decade
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Multiple value resistances with 4mm sockets
Each decade is selected by a rotary switch. Ranges cover 0 to 1111110 ohms with a resolution of 1 ohm. The tapping points are provided across each range so the unit can be used as an accurage potential divider.
All resistance values are of ±1% tolerance and of 2 watts maximum rating:
1 - 10 Ohms in steps of 1 Ohm
10 - 100 Ohms in steps of 10 Ohm
100 - 1000 Ohms in steps of 100 Ohms
1 K - 10 Kohms in steps of 1K
10 K - 100 Kohms in steps of 10 K
100 K - 1000 Kohms in steps of 100 K
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty