Would you like to log in again?
Spoke Pulley
Supplier: EISCO SCIENTIFIC LLC
SPKP14
470219-542EA 27.89 USD InStock
470219-542
Spoke Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
This pulley is ideal for use with photogates sensors. Pulley is mounted on free-running ball bearings with minimum friction. Mounted on a removeable rod of 150 x 10 mm diameter.
Diameter 65 mm with spokes
Ideal for use with Photogates
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty