Would you like to log in again?
Friction Board
Supplier: VWR International
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
PH10568
470219-584CS 168 USD InStock
470219-584
Friction Board
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
To Study the effects of friction using 4 different surface materials on a board.
  • Introduce friction
  • Includes four surface materials
  • Instructions included
Includes surface board, hooks, and wooden blocks, with instruction manual.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty