Would you like to log in again?
Complete Meter Stick Optics Bench
Supplier: United Scientific Supplies
OBSET1 OBSET2-NS
470219-700EA 36.65 USD InStock
470219-700 470219-702
Complete Meter Stick Optics Bench
Educational Materials Physics Educational Materials Optics Learning Activities
Classical experiment for studying optics on a budget.
  • All components connect to any standard meter stick
Student optical bench contains sufficient equipment for high school physics applications.
Contents:
1 Meter Stick
1 Screen support
2 lens supports for 38mm and 75mm lenses/mirrors
5 Board screens measuring 10 x 125 cm with a millimeter scale on both sides
1 Marker and object
2 Metal supports for meter stick
1 Candle holder
1 Paraffin candle
1 Activity guide
Required but not included are lenses. Focal lengths under 50cm work best.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty