Would you like to log in again?
3B Scientific® Individual Bones
Supplier: AMERICAN 3B SCIENTIFIC SE
1000139 1019365 1019369 1019363 1019359 1019362 1019370 1019373 1000137 1019375 1019364 1000143 1019360 1019367 1019366 1000142 1019355 1019371 1019357 1000140 1019358 1019368 1019377 1019372 1019376 1000136 1019374 1019356
470219-808EA 35 USD InStock
470219-808 470118-974 470118-976 470219-796 470219-830 470219-798 470219-810 470219-832 470219-794 470219-816 470219-818 470219-812 470219-834 470103-100 470219-814 470219-836 470030-152 470118-966 470118-968 470219-820 470219-804 470219-826 470219-828 470219-806 470219-822 470103-054 470219-800 470219-802
3B Scientific® Individual Bones
Models Human Biology Models
Individual Bones And Bone Sets In High Quality Plastic
  • High Quality Plastic Bones And Bone Sets
  • Use Individually, Or As Replacement Parts For A Disarticulated Skeleton
  • Includes 3B Smart Anatomy
Individual bones, and bone sets , are a great alternative to a full skeleton. Individual bones can be examined and their placement in a skeleton inferred from their structure. Use these bones and bone sets as part of an inquiry activity, as replacements for missing bones from a disarticulated skeleton, or as additional bones in a forensics activity. Unfortunately we can not guarantee selection of Left or Right side for the individual bones, but we attempt to accommodate requests when possible.
Where noted some bone sets have different articulation.
Wire: Traditional Wire Articulation, the bones are joined with a metal wire, and are limited in their flexibility. In most cases this approximates natural range of movement. When positioned, bones tend to stay in place.
Nylon: Similar to Wire Articulation, the bones are held together on a nylon line. These tend to be more loosely bound and are more flexible than wire articulation. When positioned bones may sag or move slightly.
Bungee: Bungee Articulation is much more flexible. Individual bones are held together with an elasticized cord, giving a great level of flexibility. Bones will be likely to shift after being positioned.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty