Would you like to log in again?
Science Take-Out® Breast Cancer Risk: Genes And The Environment
Supplier: Science Take-Out
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
STO-146U STO-146 STO-146-NOTG STO-146R
470219-851EA 20 USD InStock
470219-851 470219-852 470219-854 470219-850
Science Take-Out® Breast Cancer Risk: Genes And The Environment
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Explore Genetic And Environmental Risk Factors For Breast Cancer.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
An individual’s risk of developing breast cancer is influenced by a number of factors such as specific genes, lifestyle choices, hormonal factors, and environmental exposures. Analyze a family's pedigree for the occurrence of breast cancer. Conduct simulated DNA testing for the BRCA1 gene. Complete a survey on risk factors associated with breast cancer and then analyze the survey responses based on a "Breast Cancer Risks" factsheet. Analyze data from an experiment indicating that environmental exposures during childhood (before puberty) may increase risks of breast cancer. Use beads to model effects of DDT insecticide exposure before puberty and after puberty. This complete experiment setup contains all materials and does not require lab equipment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty