Would you like to log in again?
Worldwide Travel Adapter Set
Supplier: INTERNATIONAL VIDEO & ELECTRONICS
52-9410
470219-932EA 17.9 USD InStock
470219-932
Worldwide Travel Adapter Set
Power Supplies
The Worldwide Travel Adapter Set is ideal for use with most portable electronics, including notebook computers, cellular phones, MP3 players, and digital cameras. The travel plug works in over 150 countries and its compact design is ideal for travel. The universal faceplates accept virtually all types of plugs and individual pieces nest together making it easy to travel anywhere in the world.
Power Rating: 2.5A/250V max., 6A/125V max., 100–240 VAC (100VAC/600 Watt max., 230VAC/575 Watt max.)
THIS IS NOT A VOLTAGE CONVERTER OR TRANSFORMER. It does not reduce 230V to 110V.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty