Would you like to log in again?
Aerodynamics – Air Resistance, Drag Coefficient, and Pressure Curve
Supplier: KLINGER EDUCATION PROD CORP SE
P1861-2-3
470220-088KT 13788.05 USD InStock
470220-088
Aerodynamics – Air Resistance, Drag Coefficient, and Pressure Curve
Educational Materials Physics Educational Materials Mechanics Learning Activities
Precision manometer measures wind speed and pressure.
  • For Measuring Air Resistance
  • Suction and Pressure Fan
  • Saddle Base
  • Aerodynamics Working Section
  • Machine Determines Drag Coefficient
Users examine the relationship between the air resistance and the flow speed with a circular disk. The flow speed is measured with a pressure head sensor and the air resistance with a dynamometer. The machine also determines the drag coefficient for various flow bodies with equal cross-sections. The flow speed is measured using a pressure head sensor and the air resistance with a dynamometer.
Users measure the static pressure at various points on the underside of an airfoil profile. The measured curve not only illustrates the air resistance but also explains the lift acting on the airfoil.
Ordering information: This kit includes a suction and pressure fan, aerodynamics working section, aerodynamics accessories, measurement trolley for wind tunnel, sector dynamometer 0.65 N, pressure head, precision manometer, small and V-shaped base, saddle base, 47 cm and 12 mm ø stand rod, and airfoil with lateral sheets.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty