Would you like to log in again?
Electron Spin Resonance
Supplier: KLINGER EDUCATION PROD CORP SE
P6262
470220-118KT 22220.4 USD InStock
470220-118
Electron Spin Resonance
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Electron Spin Resonance determines the magnetic field as a function of resonance frequency.
  • All Major Components Included
  • Experiment Verifies Electron Spin Resonance in DPPH
  • DPPH: Diphenylpicryl-Hydrazyl
  • DPPH Is a Radical with a Free Electron Present in a Nitrogen Atom
  • Evaluation's Aim: to Determine g Factor
The experiment verifies electron spin resonance in diphenylpicryl- hydrazyl (DPPH). DPPH is a radical, in which a free electron is present in a nitrogen atom. In the experiment, the magnetic field B which fulfills the resonance condition the resonance frequencies n can be set in a continuous range from 13 to 130 MHz. The aim of the evaluation is to determine the g factor.
Ordering information: Kit Includes: ESR basic unit; ESR control unit; Pair of Helmholtz coils; BNC cable, 1 m (2); Saddle Base (3); Connecting lead 32 A, 25 cm, black; Connecting lead 32 A, 50 cm, red; and Connecting lead 32 A, 50 cm, blue. Required but not included is an Oscilloscope 30 MHz, two-channel.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty