Would you like to log in again?
X–Ray and Tomography
Supplier: KLINGER EDUCATION PROD CORP SE
P6381-2-3-4-5
470220-124KT 64794.9 USD InStock
470220-124
X–Ray and Tomography
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Five different experiments exploring X–rays and tomography.
  • Includes all major components
  • Investigate computed tomogram
In 1963 and 1964 Allan Cormack published the theoretical foundations of computer tomography in the Journal of Applied Physics. In 1972 the first computed tomographic scanner was built by Godfrey Hounsfield who, together with Allan Cormack, was awarded the Nobel Prize in Physiology or Medicine in 1979. The basic idea of computerized tomography is the illumination of an object by X-rays from numerous different angles.
Our educational X-RAY APPARATUS (preferred with W TUBE or AU TUBE) allows the illumination of objects by X-rays. The resulting 2D-projections are visualized at the fluorescence screen. These projections have a relatively low intensity. Therefore a camera of high sensitivity has to be used to record the various projections. Such a camera is implemented in the COMPUTED TOMOGRAPHY MODULE.
Each of the experiments explores a different part of X–Ray Physics:
1. Measurement and presentation of a computed tomogram with the computed tomography module
2. Computed tomography of simple geometrical objects with the computed tomography module
3. Medical basics of computed tomography with the computed tomography module
4. Determining absorption coefficients and Hounsfield units with computed tomography with the computed tomography module
5. Computed tomography of biological samples with the computed tomography module
Required but not included: PC with Windows XP/Vista/7/8/10 (x86 or x64)
X-ray apparatus
Computed tomography module
Phantom, 3D
LEGO® Adapter
Goniometer
X-ray tube Au
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty