Would you like to log in again?
Solid State Physics – Superconductivity
Supplier: KLINGER EDUCATION PROD CORP SE
P7261
470220-134KT 7021.49 USD InStock
470220-134
Solid State Physics – Superconductivity
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Determine the transition temperature of a superonductor.
  • Includes all major components
  • YBa2Cu307 suprconductor
Determine the transition temperature of the high-temperature superconductor YBa2Cu3O7‐x. For this purpose, the substance is cooled to below its critical temperature of Tc = 92 K using liquid nitrogen. In a four-point measurement setup, the voltage drop across the sample is measured as a function of the sample temperature using the computer-assisted measured value recording system CASSY.
Required but not included: PC with Windows XP/Vista/7/8/10 (x86 or x64).
Transition temperature and electrical resistance, experiment kit
Sensor-CASSY 2
CASSY Lab 2
Connecting lead 19 A, 50 cm, red/blue, pair (2)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty