Would you like to log in again?
Effect of Salinity on Seawater
Supplier: ALDON CORP SE
IS9707
470220-216KT 95.3 USD InStock
470220-216
Effect of Salinity on Seawater
Educational Materials Environmental Educational Materials Environmental Learning Activities
Water Bodies are Dynamic Systems
  • Students will visualize what happens when water with different densities mixes
The world’s ocean system contain a dynamic thermohaline current system that is responsible for many aspects of our environment. The experiments in this kit have been designed to help visualize the impact solutions of different density have on each other when mixed. Students will measure the salinity of three different water samples representing three major salt water bodies, as well as perform an experiment in which they will observe the flow patterns when different density solutions are mixed.
Ordering information: Kit contents include Phenolphthalein Indicator, Potassium Chromate, Silver Nitrate, Sodium Hydroxide, Synthetic Atlantic Ocean Water, Synthetic Gulf of Mexico Water, and Synthetic Pacific Ocean Water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty