Would you like to log in again?
Concepts of Molarity
Supplier: ALDON CORP SE
IS2519
470220-224KT 95.2 USD InStock
470220-224
Concepts of Molarity
Educational Materials Chemistry Educational Materials Periodic Table Learning Activities
Designed to Introduce Students to the Concept of Molarity.
  • Practice proper lab techniques
  • Visually determine the unknowns
Students will first try to make a solution with a specific molarity demonstrating the importance of good laboratory technique. In the second set of experiments students will perform a titration on a known acid solution before using the concepts they have learned to identify the concentrations in three unknown solutions.
Ordering information: Kit includes Hydrochloric Acid, Phenolphthalein Solution, Sodium Hydroxide, Sucrose, Unknown Hydrochloric Acids #1-#3, Teacher's Manual, and Student Study Guide Copymasters. Materials needed but not included are (15) 100 mL Beakers, (15) 200+ mL Beakers,(15-30) Burettes, (15) Burette Funnels, (15) Burette Clamps, (15) Burette Stands, Deionized Water, Graduated Cylinder, Permanent Marker, and Stirring Rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty