Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Budget Antibiotic Disks
Supplier: ALDON CORP SE
XD1398-PK/50 XD1360-DISP XD1363-PK/50 XD1362-PK/50 XD1365-PK/50 XD1364-PK/50 XD1368-PK/50 XD1360-SET XD1397-PK/50 XD1366-PK/50 XD1361-PK/50
470220-246EA 48.75 USD InStock
470220-246 470220-236 470220-244 470220-234 470220-228 470220-226 470220-238 470220-242 470220-232 470220-240 470220-230
Budget Antibiotic Disks
Culture Dishes Petri Dishes
Users can observe the sensitivity of various bacteria and fungi to an array of antibiotics with these convenient kits.
  • Excellent Quality at an Economical Cost
  • Convenient and Reliable
  • Process Demonstrates Principles of Antibiotic Reactions
  • Fifty Reseable Disks in One Sleeve
Each resalable sleeve contains 50 discs of antibiotic; all you need to do is place the disc on the bacterial culture and observe the results to determine susceptibility or resistance of an organism to specific antibiotics.
Disc dispenser, required, is available separately.
Antibiotic Sensitivity Disc Set comes with 1 magazine of each of the following: Penicillin, Streptomycin, Tetracycline, Neomycin and Erythromycin
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty