Would you like to log in again?
Complete Scintillation Probe Assembly
Supplier: SPECTRUM TECHNIQUES, LLC
SPA38
470220-454EA 1984.59 USD InStock
470220-454
Complete Scintillation Probe Assembly
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Gamma Spectroscopy detector.
  • Unit provides a complete detector assembly for gamma spectroscopy applications
The 38x38 mm NaI(Tl) crystal includes PMT and voltage divider as an integral assembly. The probe is supplied complete with a 10-position sample stand, lead shield and cables for connecting directly to the spectrometer. Resolution is typically 7% @ 662 keV.
Ordering information: Includes scintillation detector, 38x38mm with voltage divider; GMS38 position stand with sample tray and LS038 lead shield; and BNC6 and MHV6 cable assemblies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty