Would you like to log in again?
Radioactive Needle Sources
Supplier: SPECTRUM TECHNIQUES, LLC
NSSR90 NSPO210 NSSR90 NSPO210 NSPB210 NSPB210
470220-476EA 45.45 USD InStock
470220-476 470337-079 470220-474 470337-080 470337-081 470220-472
Radioactive Needle Sources
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Quick and easy way to show emission with cloud chambers.
  • Safe to use
  • Easy disposal
Needle sources are used to generate a point source of radiation inside cloud chambers for demonstrating alpha and beta radiation tracks. Three different types of isotopes are offered, a pure alpha emitter, a pure beta emitter and a combined alpha/beta emitter.
The sources are constructed by depositing a small, license exempt quantity of radioactive isotope onto the eye of a standard sewing needle which is mounted on a test tube stopper for insertion into the cloud chamber. The needle and stopper are placed into a test tube for protection during shipping and storage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty