Would you like to log in again?
Radioactive Source, Set of 3
Supplier: SPECTRUM TECHNIQUES, LLC
RSS3-UN RSS3UN RSS3
470220-510PK 212 USD InStock
470220-510 470337-068 470220-462
Radioactive Source, Set of 3
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Convenient set of 3 for studying radioactivity.
  • Safe to use
  • Easy disposal
The solid sources we supply are securely encapsulated in heat-resistant, waterproof plastic disks 25 mm in diameter and 4 to 6 mm thick. The sample is deposited in a well that is then filled with an epoxy material, safely sealing in the radioactivity.
Pulonium-210 (Po-210), Solid Activity: 0.1 µCi, Emission: α,y (half life 138 days)
Strontium-90 (Sr-90), Solid Activity: 0.1 µCi , Emission: β (half life 29 years)
Cobalt-60 (Co-60), Solid Activity 1 µCi, Emission: β,y (half life 5.3 years)
Set 470220-462 is the same except Cobalt-60 has an activity of only 0.5 µCi.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty