Would you like to log in again?
Radioactive Source, Set of 8
Supplier: SPECTRUM TECHNIQUES, LLC
RSS8 RSS8UN RSS8EU RSS8UN RSS8EU
470337-083EA 563.09 USD InStock
470337-083 470220-464 470220-514 470220-512 470337-082
Radioactive Source, Set of 8
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Convenient set of 8 for studying gamma spectroscopy.
  • Safe to use
  • Easy disposal
  • All Gamma emissions
This set contains eight different gamma emitting isotopes covering the entire energy range from 32 to 1333 keV. Also included in the set is a mixed source of Cs-137 and Zn-65 which students may use to identify an “unknown” isotope.
The set consists of Ba-133, Cd-109, Co-57, Co-60, Cs-137, Mn-54. Na-22 and Cs/Zn.
Source activities are all 1 uCi, except the Cs which is 0.5 μCi and Zn which is 1 μCi Zn.
470220-464 is the same set except the activity of Cesium is 0.25 μCi.
470220-512 is the same set except the activity of Cesium is 0.1 μCi.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty