Would you like to log in again?
Compound Pendulum
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SP394
470220-610EA 40.7 USD InStock
470220-610
Compound Pendulum
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
The compound pendulum consists of a steel bar (1 Meter long) drilled at 50mm intervals, removable knife edges and a support bracket for mounting on walls.
  • Sturdy Design
  • Repeatable results
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty