Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Animal Fur

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

WARD470220-908
470220-910PK 76.3 USD InStock
470220-910 470220-914 470220-912 470220-908
Animal Fur
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This real animal fur is for use in electrostatic experiments.

  • Real Fur Produces Better Charges than Fake Can Produce
  • Pelt's Length Ranges from 9 to 6"
  • Pelt's Width Ranges from 16 to 12"
  • Average Pelt-Strip Length: 5"
  • Average Pelt-Strip Width: 6"

Traditional methods of creating a negative charge involve a rubber rod and fur, which are still best for investigating electrostatics in the classroom.

Pelts range in size from approximately 9 x 6" to 16 x 12". No size choices are available. Pelt strips range in size, but the average size is 5 x 6".

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty