Would you like to log in again?
Rudiger® Physiology Skeleton
Supplier: Rudiger
A203
470221-120EA 976.7 USD InStock
470221-120
Rudiger® Physiology Skeleton
Educational Materials Biology Educational Materials Human Biology Learning Activities
Natural motion in every part: upper and lower limb, hand, feet, head, shoulder, hip, and spine.
  • Superior flexibility
  • Integrated bone movement
  • 5 year manufacturers warranty
Medial and lateral rotation of arms and legs, flexion and extension of elbows and knees. Shoulders are flexibly mounted to show natural hyperextension, pronation and supination of the forearms. One hand and foot mounted on elastic cord. Flexible spine with extra soft discs to show life-like deformation while stretching and bending spine. With spinal cord, nerve ends, arteries, sectioned sacrum, sacral flap, 5-legs safety stand and dust-cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty