Would you like to log in again?
Rudiger® Physiological Spine
Supplier: Rudiger
A207
470221-152EA 294.65 USD InStock
470221-152
Rudiger® Physiological Spine
Educational Materials Biology Educational Materials Human Biology Learning Activities
Lifelike accuracy cast from a natural spine.
  • Fully articulated and flexible
  • Soft discs compress like natural tissue
  • 5 year manufacturers warranty
Our spine is fully articulated and flexible to show vertebral movements and alignments. Comes with arteries, spinal cord, nerve ends and flexible femur heads. The sacrum is sectioned to show course of spinal nerves. Contains a softer disc material, that compresses like natural tissue, bulging outward when under pressure. With stand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty