Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Conical Tube, Natural, Sterile, Bulk Packaged, 50 ml, Heathrow Scientific®

Supplier: Heathrow Scientific
HS4427B
470221-264CS 317.9 USD InStock
470221-264
Conical Tube, Natural, Sterile, Bulk Packaged, 50 ml, Heathrow Scientific®
Tubes Test Tubes
Sterile, bulk packaged, natural colored 50 ml conical tubes.

  • Lot-to-lot traceability with easy-tear packaging
  • Maximum RCF rating is 9500 ×g
  • Graduations provided in 5.0 ml increments
  • Writing area and flat polyethyelene lids for easy labeling

Imprinted graduations and a self-standing version (only in clear bulk packs).

Packaging: Tubes are packaged 25 per bag, 20 bags per case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty