Would you like to log in again?
Nanotechnology
Supplier: THAMES & KOSMOS
631727
470221-496EA 123.65 USD InStock
470221-496
Nanotechnology
Educational Materials Physics Educational Materials Applied Physics Learning Activities
With this kit, the intangible, nanoscopic world becomes accessible with hands-on experiments using both scaled-up models and real nanomaterials.
  • Instructions Included
  • Kit Features a Variety of Experiments
  • Cutting-Edge Nanomaterials
  • Students Conduct Experiments Concerning Size, Scale, and Orders of Magnitude
  • Subjects Include Laser Light, Electron Microscopy, and More
Nanoparticles can be made of different materials and exist in a wide variety of forms. These particles can be coated onto surfaces, enhance other materials such as paint, or used as a delivery method for pharmaceuticals in the body. Some applications include self-cleaning house paints and water-repellent clothes, anti-fog coatings on glasses, UV protection components in sunscreen, environmentally friendly antifouling coatings for ships (to prevent algae growth, thereby reducing fuel consumption), antibacterial coatings for medical devices, and materials to more effectively filter dirty water.
Students begin by conducting experiments concerning size, scale, and orders of magnitude in the International System of Units to develop an understanding of how small a nanometer is. With filtering experiments, they can explore how charcoal is similar to a nanoscale sponge. They can also conduct experiments with the hydrophilic (water-loving) and hydrophobic (water-hating) properties of different surfaces, which illustrate nanoscale properties. Other activities include stirring up suspensions to examine exotic mixtures and study the nanoscopic particles suspended in them. Students learn about the electromagnetic spectrum, laser light, electron microscopy, and atomic force microscopy.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty