Would you like to log in again?
Physics Workshop Pro
Supplier: THAMES & KOSMOS
625314
470221-518EA 119.2 USD InStock
470221-518
Physics Workshop Pro
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Physics Workshop Pro explores advanced subjects in physics.
  • Ninety-Six-Page, Illustrated Instruction Manual
  • Topics Include Fluid Dynamics, Energy, Oscillation, and More
  • Buildable Models Include a Wind Tunnel, Pneumatic Shocks, and a Hydraulic Lift
  • Kit Features More than 212 Parts
  • Kit's Main Focus: Water and Air
These topics include fluid dynamics, energy, oscillation, hydraulics, and pneumatics. With a grasp of these advanced topics, students can build spectacular models and devices such as a wind tunnel, pneumatic shocks, and a hydraulic lift. This workshop shows students how physics extends beyond the classroom to give us an amazing set of tools that can be applied to engineer marvelous things. The main focus of the kit is the behavior of the two most important fluids in our world, water and air, and objects immersed in them. Students investigate how air and water rest and flow, what they can carry, and how they move. With experiments and models, students learn how ships float and airplanes fly, why a streamlined shape can make a car drive fast, and how power plants convert a current of water into electrical current.
The 96-page, full-color experiment manual has two major sections. The first sections describes the properties of air, water, currents, and energy. It features 17 experiments with instructions for building smaller models. The second section includes instructions for building 14 larger models of complex devices. This kit features more than 212 parts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty