Would you like to log in again?
Photosynthesis and Respiration Chamber Demonstration
Supplier: ALDON CORP SE
IS6501
470221-560EA 42.6 USD InStock
470221-560
Photosynthesis and Respiration Chamber Demonstration
Educational Classroom Kits and Activities
How are Photosynthesis and Respiration Related?
  • Discover the correlation between Oxygen and Carbon Dioxide levels in the chamber
  • Examine how light intensity affects these levels
The photosynthesis/respiration demonstration chamber may be used to monitor oxygen levels, carbon dioxide levels, or both in an enclosed environment. The chamber has been designed to be used with a Oxygen logger sensor and/or a Carbon dioxide logger sensor as well as software for data acquisition. As an option, a light sensor may also be used when running photosynthesis experiments to correlate light intensity to changes in carbon dioxide and oxygen levels in the chamber.
The lid of the chamber provides openings for both the oxygen sensor and the carbon dioxide sensor. If both sensors will not be used for a particular experiment, place the proper rubber stopper in the opening that will not contain a sensor during the experimental run. You can set up an experiment and record light and dark cycles over a weekend or period of time without even being in the classroom as the sensors will capture all of the information for your student to study. Required: O2, CO2, Light sensors
Materials Included:
Clear glass bottle with cap
Rubber stoppers
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty