Would you like to log in again?
Effectiveness of Sunscreen
Supplier: ALDON CORP SE
IS6502
470221-562EA 42.6 USD InStock
470221-562
Effectiveness of Sunscreen
Educational Materials Environmental Educational Materials Environmental Learning Activities
This activity illustrates the importance sunscreen use.
  • For Use with NeuLog Logger Sensors
  • Students See the Importance of Using Sunscreen
  • Kit Provides Microscope Slides and SPF 15 and 30
  • Teacher's Guide and Student Copy Master Included
Electromagnetic radiation in the UV range of the spectrum cannot be perceived by the human eye. Using the NeuLog UVB sensor specifically designed to measure UVB radiation, users compare the amount of UVB radiation reaching the sensor.
The sensor is covered by both SPF 15 and SPF 30 sunscreens to determine if the sunscreens actually provide UVB protection, and if so, how much protection. 
Ordering information: This kit includes SPF 15 and SPF 30, microscope slides, a teacher's guide, and a student copy master. A NeuLog Logger and NeuLog UVA and UVB sensors are required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty