Would you like to log in again?
Disappearing Rainbow Demonstration
Supplier: ALDON CORP SE
IS7039
470221-810EA 41.6 USD InStock
470221-810
Disappearing Rainbow Demonstration
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Demonstrate the concepts of Indicators and Acids and Bases!
  • Students see reactions in real time
  • Combine colors to create all the colors of the rainbow
Three indicators are used in conjunction with acids and bases to create a rainbow of colors. In a basic solution, phenolphthalein is red, p-nitrophenol is yellow, and thymolphthalein is blue. If done correctly the students only see the acid or base being poured, which adds to the impressive display. There are enough materials to complete the demonstration 7 times.
Ordering information: Materials include Red Indicator, Orange Indicator, Green Indicator, Blue Indicator, Violet Indicator, Hydrochloric Acid 1.0N, Sodium Hydroxide 1.0N, Sodium Hydroxide 6.0M, Instruction Manual. Material Needed but not included: (6) Glass Beakers or Clear Plastic Cups (at least 250mL), Graduated Cylinder, 100mL Distilled or Deionized water. Materials needed but not included are (6) Glass beakers or clear plastic cups (at least 250ml), graduated cylinder, 100ml, distilled or deionized water.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty